missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Desloratadine, TRC

Product Code. 30004672
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004672 500 mg 500mg
17904415 250 mg 250mg
17993615 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004672 Supplier Toronto Research Chemicals Supplier No. TRCM316500500MG
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38092-89-6
Molecular Formula C20 H21 Cl N2
Molecular Weight (g/mol) 324.85
Synonym 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine, Loratadine Imp. G (EP)
IUPAC Name 8-chloro-11-(1-methylpiperidin-4-ylidene)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridine
SMILES CN1CCC(=C2c3ccc(Cl)cc3CCc4cccnc24)CC1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C20H21ClN2/c1-23-11-8-14(9-12-23)19-18-7-6-17(21)13-16(18)5-4-15-3-2-10-22-20(15)19/h2-3,6-7,10,13H,4-5,8-9,11-12H2,1H3
Formula Weight 324.1393
Percent Purity >95%
Chemical Name or Material 8-Chloro-11-(1-methylpiperidin-4-ylidene)-6,11-dihydro-5H-benzo[5,6]cyclohepta[1,2-b]pyridine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.