missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-Methyl-D-glucamine, 99%

Product Code. 10123560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123560 500 g Plastic bottle 500g
10783881 100 g Plastic bottle 100g
10043993 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10123560 Supplier Thermo Scientific Chemicals Supplier No. 126845000
  • ONLINE EXCLUSIVE PRICE
    £80.30
    £72.65 / 500g
    Save £7.65
    9% Off
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

N-Methyl-D-glucamine, CAS # 6284-40-8, also known as meglumine, is a base compound commonly used in combination with iodinated organic compounds in contrast medium. | CAS: 6284-40-8 | C7H17NO5 | 195.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information


• N-Methyl-D-glucamine is a hexosamine compound that is frequently used in combination with iodinated organic compounds in contrast media such as iothalamate meglumine, iodipamide meglumine, and diatrizoate meglumine


Applications


• It is also useful when applied in ion-exchange resins to chelate boron ions selectively
• It is considered a base and frequently used in the preparation of salts of certain acids
• It is also applied in the synthesis of surface-active agents and laboratory dyes

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6284-40-8
Molecular Formula C7H17NO5
Molecular Weight (g/mol) 195.21
MDL Number MFCD00004707
InChI Key MBBZMMPHUWSWHV-BDVNFPICSA-N
Synonym meglumine, n-methyl-d-glucamine, n-methylglucamine, meglumin, 1-deoxy-1-methylamino-d-glucitol, methylglucamin, meglumina, 1-deoxy-1-methylaminosorbitol, d-glucitol, 1-deoxy-1-methylamino, n-methyl-d--glucamine
PubChem CID 8567
ChEBI CHEBI:59732
IUPAC Name (2R,3R,4R,5S)-6-(methylamino)hexane-1,2,3,4,5-pentol
SMILES CNCC(C(C(C(CO)O)O)O)O

Specifications

Absorbance 0.03 max. (20 w/v% aq. soln.) at 420nm
CAS Min % 98.5
CAS Max % 100.0
Melting Point 128.0°C to 132.0°C
Color White
Loss on Drying 1% max. (1 g, 100°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Beilstein 04, IV, 1914
Merck Index 15, 6150
Specific Rotation Condition − 16.50 (20.00°C c=10,H2O,on dry sub)
Specific Rotation − 16.50
Solubility Information Solubility in water: 100 g/100 mL water (25°C). Other solubilities: 1.2 g/100 mL alcohol (25°C), 21 g/100 mL alcohol (70°C)
Formula Weight 195.21
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-D-glucamine, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.