missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride (Secondary Amine Hydrochloride), Mikromol™

Product Code. 17943766
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943766 25 mg 25mg
17922146 100 mg 100mg
30458756 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17943766 Supplier Mikromol Supplier No. MM0696.020025
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride (Secondary Amine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride
CAS 65473-13-4
Analyte or Component Names N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C12 H13 N . Cl H
Certifications/Compliance ISO 17025
Synonym Terbinafine Hydrochloride Imp. A (EP), Terbinafine Imp. A (EP), Terbinafine USP Related Compound A, Terbinafine USP RC A, N-Methyl-C-(naphthalen-1-yl)methanamine Hydrochloride, Terbinafine Hydrochloride Imp. A (EP) as Hydrochloride, Secondary Amine Hydrochloride of Terbinafine, Terbinafine Related Compound A as Hydrochloride, Terbinafine Hydrochloride Impurity A as Hydrochloride, Terbinafine Impurity A as Hydrochloride
InChI Formula InChI=1S/C12H13N.ClH/c1-13-9-11-7-4-6-10-5-2-3-8-12(10)11;/h2-8,13H,9H2,1H3;1H
SMILES Cl.C=1C=CC2=C(C1)C=CC=C2CNC
IUPAC Name N-methyl-1-naphthalen-1-ylmethanamine;hydrochloride
Molecular Weight (g/mol) 207.7
Quantity 25 mg
Formula Weight 207.0815 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.