Learn More
N-Methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine, 97%, Maybridge™
Brand: Maybridge CC58146DA
Code : 31
Additional Details : CAS Number : 886851-54-3 Weight : 0.05100kg Transport : UN number : 2735 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
886851-54-3 | |
264.137 | |
FJHBRYOEMIQSPC-UHFFFAOYSA-N | |
18525892 | |
CNCC1=C2C(=C(S1)Br)OCCO2 |
C8H10BrNO2S | |
MFCD08690308 | |
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine | |
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine |
Specifications
N-methyl-(7-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-5-yl)methylamine | |
1g | |
89.5 | |
90% | |
MFCD08690308 | |
FJHBRYOEMIQSPC-UHFFFAOYSA-N | |
1-(5-bromo-2,3-dihydrothieno[3,4-b][1,4]dioxin-7-yl)-N-methylmethanamine | |
18525892 | |
90% |
Amber Glass Bottle | |
886851-54-3 | |
100.0 | |
C8H10BrNO2S | |
n-methyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl methylamine, n-methyl-5-aminomethyl-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxine, 7-bromo-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl methyl methyl amine, 7-bromo 2h,3h-thiopheno 4,3-e 1,4-dioxan-5-yl methyl methylamine, thieno 3,4-b-1,4-dioxin-5-methanamine,7-bromo-2,3-dihydro-n-methyl, 1-7-bromo-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-n-methylmethanamine | |
CNCC1=C2C(=C(S1)Br)OCCO2 | |
264.137 | |
264.14 |