missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine, 97%, Thermo Scientific™

Product Code. 10156663
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10156663 1 g Amber Glass Bottle 1g
10644205 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10156663 Supplier Thermo Scientific Maybridge Supplier No. CC58346DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-99-6
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.288
MDL Number MFCD09879908
InChI Key CWRQCHDTRDIGCU-UHFFFAOYSA-N
Synonym n-methyl-6-tetrahydropyran-4-yloxy pyrid-3-yl methylamine, 5-methylamino methyl-2-tetrahydro-2h-pyran-4-yloxy pyridine, methyl 6-oxan-4-yloxy pyridin-3-yl methyl amine, n-methyl-1-6-oxan-4-yl oxy pyridin-3-yl methanamine, methyl 6-2h-3,4,5,6-tetrahydropyran-4-yloxy 3-pyridyl methyl amine, n-methyl-1-6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl methanamine
PubChem CID 24229584
IUPAC Name N-methyl-1-[6-(oxan-4-yloxy)pyridin-3-yl]methanamine
SMILES CNCC1=CN=C(C=C1)OC2CCOCC2

Especificaciones

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material N-Methyl-[6-(tetrahydropyran-4-yloxy)pyrid-3-yl]methylamine
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.