missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-5-tetrazolethiol, TRC

Product Code. 30133289
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133289 100 mg 100mg
30004937 1 g 1g
30010553 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133289 Supplier Toronto Research Chemicals Supplier No. TRCM330260100MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13183-79-4
Molecular Formula C2 H4 N4 S
Molecular Weight (g/mol) 116.145
Synonym 1-Methyl-1H-tetrazole-5-thiol, Cefoperazone Sodium Imp. C (EP), 5H-Tetrazole-5-thione, 1,2-dihydro-1-methyl- (9CI, ACI), 1,2,3,4-Tetrazole-5-thiol, 1-methyl- (4CI), 1,2-Dihydro-1-methyl-5H-tetrazole-5-thione (ACI), 1H-Tetrazole-5-thiol, 1-methyl- (7CI, 8CI), 2-Tetrazoline-5-thione, 1-methyl- (6CI), 1-Methyl-1,2,3,4-tetrazole-5-thiol, 1-Methyl-1,2,3,4-tetrazoline-5-thione, 1-Methyl-1H-1,2,3,4-tetrazole-5-thiol, 1-Methyl-1H-tetrazol-5-thiol, 1-Methyl-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione, 1-Methyl-2H-tetrazole-5-thione, 1-Methyl-5-mercapto-1,2,3,4-tetrazole, 1-Methyl-5-mercapto-1H-tetrazole, 1-Methyl-5-mercaptotetrazole, 1-Methyl-5-tetrazolethione, 1-Methyl-5-tetrazolinethione, 1-Methyl-5-thiotetrazole, 1-Methyltetrazole-5-thiol, 5-Mercapto-1-methyl-1,2,3,4-tetrazole, 5-Mercapto-1-methyl-1H-tetrazole, 5-Mercapto-1-methyltetrazole, [1-Methyl-1H-1H-tetrazol-5-yl]thiol, N-Methyl-5-tetrazolethiol, NSC 520787, T 1982F
IUPAC Name 1-methyltetrazole-5-thiol
SMILES Cn1nnnc1S

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C2H4N4S/c1-6-2(7)3-4-5-6/h1H3,(H,3,5,7)
Formula Weight 116.0157
Percent Purity >95%
Chemical Name or Material 1-Methyl-1H-tetrazole-5-thiol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.