missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(5-phenylpyrid-2-yl)methylamine 97%, Thermo Scientific™

Product Code. 10113774
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10113774 1 g Amber Glass Bottle 1g
10154054 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113774 Supplier Thermo Scientific Maybridge Supplier No. CC55046DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-02-2
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
MDL Number MFCD10700051
InChI Key HHOWXRCPAWWJPH-UHFFFAOYSA-N
Synonym n-methyl 5-phenylpyrid-2-yl methylamine, methyl 5-phenylpyridin-2-yl methyl amine, 2-methyl aminomethyl-5-phenylpyridine, 2-pyridinemethanamine,n-methyl-5-phenyl, methyl 5-phenyl 2-pyridyl methyl amine, n-methyl-5-phenylpyridin-2-yl methylamine, n-methyl-1-5-phenylpyridin-2-yl methanamine
PubChem CID 26343606
IUPAC Name N-methyl-1-(5-phenylpyridin-2-yl)methanamine
SMILES CNCC1=NC=C(C=C1)C2=CC=CC=C2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 198.27
Percent Purity 90%
Chemical Name or Material n-methyl(5-phenylpyrid-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.