missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(5-bromopyrid-2-yl)methylamine, ≥95%, Thermo Scientific™

Product Code. 10402444
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10402444 1 g Amber Glass Bottle 1g
10275364 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10402444 Supplier Thermo Scientific Maybridge Supplier No. MO07896DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 915707-70-9
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.067
MDL Number MFCD09065029
InChI Key DCQLMECRAAGXSD-UHFFFAOYSA-N
Synonym n-methyl-5-bromopyrid-2-yl methylamine, 5-bromopyridin-2-yl methyl methyl amine, 5-bromo-2-methyl aminomethyl pyridine, 1-5-bromopyridin-2-yl-n-methylmethylamine, 1-5-bromopyridin-2-yl-n-methylmethanamine, 2-pyridinemethanamine,5-bromo-n-methyl, 5-bromo 2-pyridyl methyl methylamine, 2-pyridinemethanamine, 5-bromo-n-methyl, 5-bromo-pyridin-2-ylmethyl-methyl-amine
PubChem CID 24229777
IUPAC Name 1-(5-bromopyridin-2-yl)-N-methylmethanamine
SMILES CNCC1=NC=C(C=C1)Br

Specifications

CAS Min % 95.0
Assay Percent Range 95+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 201.07
Percent Purity 95%
Chemical Name or Material N-methyl-(5-bromopyrid-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.