missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-pyrimidin-2-ylbenzylamine, 90%, Thermo Scientific™

Product Code. 10451514
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10451514 250 mg Amber Glass Bottle 250mg
10002164 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10451514 Supplier Thermo Scientific Maybridge Supplier No. CC56246CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-48-5
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD09702382
InChI Key DIWVUAYHEYROLA-UHFFFAOYSA-N
Synonym n-methyl-4-pyrimidin-2-ylbenzylamine, n-methyl-1-4-pyrimidin-2-yl phenyl methanamine, methyl 4-pyrimidin-2-yl phenyl methyl amine, n-methyl-4-pyrimidin-2-yl benzylamine, benzenemethanamine,n-methyl-4-2-pyrimidinyl, methyl 4-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-4-2-pyrimidinyl-9ci
PubChem CID 24229561
IUPAC Name N-methyl-1-(4-pyrimidin-2-ylphenyl)methanamine
SMILES CNCC1=CC=C(C=C1)C2=NC=CC=N2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.26
Percent Purity 90%
Chemical Name or Material N-methyl-4-pyrimidin-2-ylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.