missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-methylbenzamide, 97%, Thermo Scientific™

Product Code. 15442928
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantity Dimensione della confezione
15442928 250 mg 250mg
15432928 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 15442928 Fornitore Thermo Scientific Alfa Aesar N. del fornitore H56446.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 18370-11-1
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.193
MDL Number MFCD00043627
InChI Key FZIOOTTWDRFBKU-UHFFFAOYSA-N
Synonym n-methyl-p-toluamide, n,p-dimethylbenzamide, n-methyl 4-methylphenyl carboxamide, n-methyl-4-toluamide, n,4-dimethyl-benzamide, n-methyl-p-methylbenzamide, 4-methyl-n-methylbenzamide, n-methyl 4-methylbenzamide, n-methyl-4-methylbenzamide
PubChem CID 140377
IUPAC Name N,4-dimethylbenzamide
SMILES CC1=CC=C(C=C1)C(=O)NC

Specifica

Quantity 250 mg
Formula Weight 149.18
Percent Purity 97%
Chemical Name or Material N-Methyl-4-methylbenzamide
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.