missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride, Tech., Thermo Scientific™

Product Code. 10215414
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215414 5 g Amber Glass Bottle 5g
10613625 250 mg Amber Glass Bottle 250mg
10672855 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215414 Supplier Thermo Scientific Maybridge Supplier No. CC54296DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-82-2
Molecular Formula C6H9BrClNS
Molecular Weight (g/mol) 242.559
MDL Number MFCD09817483
InChI Key GORGMQYTNXIOBQ-UHFFFAOYSA-N
Synonym n-methyl-4-bromothien-3-yl methyl amine hydrochloride, 1-4-bromothiophen-3-yl-n-methylmethanamine hydrochloride, 4-bromothiophen-3-yl methyl methyl amine hydrochloride, 4-bromo 3-thienyl methyl methylamine, chloride, 4-bromo-thiophen-3-ylmethyl-methyl-amine hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride tech, 1-4-bromothiophen-3-yl-n-methylmethanamine-hydrogen chloride 1/1
PubChem CID 24229540
IUPAC Name 1-(4-bromothiophen-3-yl)-N-methylmethanamine;hydrochloride
SMILES CNCC1=CSC=C1Br.Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 242.57
Percent Purity Tech.
Grade Technical
Chemical Name or Material N-methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.