missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride, Tech., Thermo Scientific™

Product Code. p-3586866
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10613625

Brand: Thermo Scientific Maybridge CC54296CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-82-2
Molecular Formula C6H9BrClNS
Molecular Weight (g/mol) 242.559
MDL Number MFCD09817483
InChI Key GORGMQYTNXIOBQ-UHFFFAOYSA-N
Synonym n-methyl-4-bromothien-3-yl methyl amine hydrochloride, 1-4-bromothiophen-3-yl-n-methylmethanamine hydrochloride, 4-bromothiophen-3-yl methyl methyl amine hydrochloride, 4-bromo 3-thienyl methyl methylamine, chloride, 4-bromo-thiophen-3-ylmethyl-methyl-amine hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride, 3-bromo-4-methylamino methyl thiophene hydrochloride tech, 1-4-bromothiophen-3-yl-n-methylmethanamine-hydrogen chloride 1/1
PubChem CID 24229540
IUPAC Name 1-(4-bromothiophen-3-yl)-N-methylmethanamine;hydrochloride
SMILES CNCC1=CSC=C1Br.Cl

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 242.57
Percent Purity Tech.
Grade Technical
Chemical Name or Material N-methyl-[(4-bromothien-3-yl)methyl]amine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.