missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(5-methyl-1,3,4-oxadiazol-2-yl)benzylamine, 95%, Thermo Scientific™

Product Code. 10143474
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143474 1 g Amber Glass Bottle 1g
10377204 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143474 Supplier Thermo Scientific Maybridge Supplier No. CC61746DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 944450-83-3
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD11109326
InChI Key PBJZWGQLXKZEJT-UHFFFAOYSA-N
Synonym n-methyl-4-5-methyl-1,3,4-oxadiazol-2-yl benzylamine, methyl 4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methyl amine, 2-4-methyl aminomethyl phenyl-5-methyl-1,3,4-oxadiazole, methyl 4-5-methyl 1,3,4-oxadiazol-2-yl phenyl methyl amine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methanamine, n-methyl-1-4-5-methyl-1,3,4-oxadiazol-2-yl phenyl methylamine
PubChem CID 26343642
IUPAC Name N-methyl-1-[4-(5-methyl-1,3,4-oxadiazol-2-yl)phenyl]methanamine
SMILES CC1=NN=C(O1)C2=CC=C(C=C2)CNC

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl-4-(5-methyl-1, 3, 4-oxadiazol-2-yl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.