missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 11350687
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11350687 250 mg 250mg
10693425 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350687 Supplier Thermo Scientific Maybridge Supplier No. CC50946CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 884507-32-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD09025868
InChI Key MUCOLQGVWQXQMX-UHFFFAOYSA-N
Synonym n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
PubChem CID 18525856
IUPAC Name N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
SMILES CNCC1=CC=C(C=C1)C1=NC(C)=NO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 250 mg
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl-[4-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.