missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-(trifluoromethyl)benzylamine, 95%

Product Code. 15470839
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470839 1 g 1g
15480839 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470839 Supplier Thermo Scientific Alfa Aesar Supplier No. H63888.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90390-07-1 | C9H10F3N | 189.181 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90390-07-1
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD04115410
InChI Key JFLPPELZYKHKQZ-UHFFFAOYSA-N
Synonym n-methyl-3-trifluoromethyl benzylamine, n-methyl-n-3-trifluoromethyl benzyl amine, n-methyl-1-3-trifluoromethyl phenyl methanamine, methyl 3-trifluoromethyl phenyl methyl amine, benzenemethanamine, n-methyl-3-trifluoromethyl, n-methyl 3-trifluoromethyl phenyl methanamine, n-methyl-1-3-trifluoromethyl phenyl methanamine hydrochloride, chembrdg-bb 4004318, methyl-3-trifluoromethyl-benzyl-amine
PubChem CID 485414
IUPAC Name N-methyl-1-[3-(trifluoromethyl)phenyl]methanamine
SMILES CNCC1=CC(=CC=C1)C(F)(F)F

Specifications

Color Colorless
Quantity 1 g
UN Number UN2735
Formula Weight 189.18
Percent Purity 95%
Physical Form Oil
Chemical Name or Material N-Methyl-3-(trifluoromethyl)benzylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.