missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-pyrimidin-2-ylbenzylamine, 97%, Thermo Scientific™

Product Code. 10357014
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-49-6
Molecular Formula C12H13N3
Molecular Weight (g/mol) 199.257
MDL Number MFCD09702386
InChI Key AHOVMAYXXRVWIS-UHFFFAOYSA-N
Synonym n-methyl-3-pyrimidin-2-ylbenzylamine, methyl 3-pyrimidin-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-pyrimidinyl, n-methyl-1-3-pyrimidin-2-yl phenyl methanamine, n-3-pyrimidin-2-yl benzyl methylamine, 2-3-methylamino methyl phenyl pyrimidine, methyl 3-pyrimidin-2-ylphenyl methyl amine, benzenemethanamine, n-methyl-3-2-pyrimidinyl-9ci
PubChem CID 24229568
IUPAC Name N-methyl-1-(3-pyrimidin-2-ylphenyl)methanamine
SMILES CNCC1=CC=CC(=C1)C2=NC=CC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.26
Percent Purity 97%
Chemical Name or Material N-methyl-3-pyrimidin-2-ylbenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'