missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-phenoxybenzylamine, 97%, Thermo Scientific™

Product Code. 10762295
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10762295 1 g 1g
10225864 250 mg 250mg
10002324 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10762295 Supplier Thermo Scientific Maybridge Supplier No. CC57146DA
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 129535-78-0
Molecular Formula C14H15NO
Molecular Weight (g/mol) 213.28
MDL Number MFCD02089413
InChI Key GDVLOOQWWBPGSV-UHFFFAOYSA-N
Synonym n-methyl-3-phenoxybenzylamine, n-methyl-1-3-phenoxyphenyl methanamine, methyl 3-phenoxyphenyl methyl amine, n-methyl-3-phenoxybenzenemethanamine, benzenemethanamine, n-methyl-3-phenoxy, acmc-209bgx, benzenemethanamine,n-methyl-3-phenoxy
PubChem CID 10512764
IUPAC Name N-methyl-1-(3-phenoxyphenyl)methanamine
SMILES CNCC1=CC(=CC=C1)OC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 213.28
Percent Purity 97%
Chemical Name or Material N-Methyl-3-phenoxybenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.