missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-nitrobenzylamine, 95%, Thermo Scientific™

Product Code. 15480929
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480929 1 g 1g
15490929 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480929 Supplier Thermo Scientific Alfa Aesar Supplier No. H63942.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Methyl-3-nitrobenzylamine is employed as a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19499-61-7
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD04115408
InChI Key NTPAPKLGADEFAM-UHFFFAOYSA-N
Synonym n-methyl-1-3-nitrophenyl methanamine, n-methyl-n-3-nitrobenzyl amine, methyl 3-nitrophenyl methyl amine, benzenemethanamine, n-methyl-3-nitro, 3-nitro-n-methylbenzylamine, n-methyl-3-nitrobenzylamine, n-3-nitrophenylmethyl methylamine, n-methyl-1-3-nitrophenyl methylamine
PubChem CID 409439
SMILES CNCC1=CC(=CC=C1)[N+](=O)[O-]

Specifications

Quantity 1 g
UN Number UN2735
Solubility Information Slightly soluble in water.
Formula Weight 166.18
Percent Purity 95%
Physical Form Oil
Chemical Name or Material N-Methyl-3-nitrobenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.