missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(3-methylisoxazol-5-ylmethyl)amine, 97%, Thermo Scientific™

Catalog Number p-3586860
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
401647-22-1
C6H10N2O
126.16
MFCD08060065
UQKWKRSUELRKSK-UHFFFAOYSA-N
n-methyl-1-3-methylisoxazol-5-yl methanamine, n-methyl-3-methylisoxazol-5-ylmethyl amine, methyl 3-methyl-1,2-oxazol-5-yl methyl amine, n-methyl 3-methylisoxazol-5-yl methanamine, methyl 3-methylisoxazol-5-yl methyl amine, n-methyl-1-3-methyl-1,2-oxazol-5-yl methanamine, 5-isoxazolemethanamine,n,3-dimethyl, 5-isoxazolemethanamine, n,3-dimethyl, methyl 3-methylisoxazol-5-ylmethyl amine, n-methyl-n-3-methylisoxazol-5-yl methyl amine
25220779
N-methyl-1-(3-methyl-1,2-oxazol-5-yl)methanamine
CNCC1=CC(C)=NO1
This item is not returnable. View return policy
401647-22-1
C6H10N2O
126.16
MFCD08060065
UQKWKRSUELRKSK-UHFFFAOYSA-N
n-methyl-1-3-methylisoxazol-5-yl methanamine, n-methyl-3-methylisoxazol-5-ylmethyl amine, methyl 3-methyl-1,2-oxazol-5-yl methyl amine, n-methyl 3-methylisoxazol-5-yl methanamine, methyl 3-methylisoxazol-5-yl methyl amine, n-methyl-1-3-methyl-1,2-oxazol-5-yl methanamine, 5-isoxazolemethanamine,n,3-dimethyl, 5-isoxazolemethanamine, n,3-dimethyl, methyl 3-methylisoxazol-5-ylmethyl amine, n-methyl-n-3-methylisoxazol-5-yl methyl amine
25220779
N-methyl-1-(3-methyl-1,2-oxazol-5-yl)methanamine
CNCC1=CC(C)=NO1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 126.16
Percent Purity 97%
Chemical Name or Material N-methyl-(3-methylisoxazol-5-ylmethyl)amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'