missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(2-thiomorpholinopyrid-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 11380677
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11380677 250 mg Amber Glass Bottle 250mg
10624015 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380677 Supplier Thermo Scientific Maybridge Supplier No. CC50546CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-66-7
Molecular Formula C11H17N3S
Molecular Weight (g/mol) 223.338
MDL Number MFCD09702364
InChI Key KEXCZGSQJYTNRX-UHFFFAOYSA-N
Synonym n-methyl-2-thiomorpholinopyrid-4-yl methylamine, n-methyl-2-thiomorpholinopyridin-4-yl methylamine, methyl 2-thiomorpholin-4-yl pyridin-4-yl methyl amine, n-methyl-1-2-thiomorpholinopyridin-4-yl methanamine, n-methyl-1-2-thiomorpholin-4-yl pyridin-4-yl methanamine, methyl 2-1,4-thiazaperhydroin-4-yl 4-pyridyl methyl amine
PubChem CID 24229511
IUPAC Name N-methyl-1-(2-thiomorpholin-4-ylpyridin-4-yl)methanamine
SMILES CNCC1=CC(=NC=C1)N2CCSCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.34
Percent Purity 97%
Chemical Name or Material N-methyl-(2-thiomorpholinopyrid-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.