missing translation for 'onlineSavingsMsg'
Learn More

n-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine, 90%, Thermo Scientific™

Catalog Number 10431884
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
921939-14-2
C10H11N3S
205.279
MFCD09879951
VZHVJAYQGDCZAB-UHFFFAOYSA-N
n-methyl-2-thien-2-ylpyrimidin-5-yl methylamine, methyl 2-thiophen-2-yl pyrimidin-5-yl methyl amine, methyl 2-2-thienyl pyrimidin-5-yl methyl amine, n-methyl-1-2-thiophen-2-yl pyrimidin-5-yl methanamine, n-methyl-2-thiophen-2-ylpyrimidin-5-yl methylamine
24229733
N-methyl-1-(2-thiophen-2-ylpyrimidin-5-yl)methanamine
CNCC1=CN=C(N=C1)C2=CC=CS2
This item is not returnable. View return policy
921939-14-2
C10H11N3S
205.279
MFCD09879951
VZHVJAYQGDCZAB-UHFFFAOYSA-N
n-methyl-2-thien-2-ylpyrimidin-5-yl methylamine, methyl 2-thiophen-2-yl pyrimidin-5-yl methyl amine, methyl 2-2-thienyl pyrimidin-5-yl methyl amine, n-methyl-1-2-thiophen-2-yl pyrimidin-5-yl methanamine, n-methyl-2-thiophen-2-ylpyrimidin-5-yl methylamine
24229733
N-methyl-1-(2-thiophen-2-ylpyrimidin-5-yl)methanamine
CNCC1=CN=C(N=C1)C2=CC=CS2
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 205.28
Percent Purity 90%
Chemical Name or Material N-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'