missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine, 95%, Thermo Scientific™

Product Code. 10174004
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10174004 1 g Amber Glass Bottle 1g
11320697 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174004 Supplier Thermo Scientific Maybridge Supplier No. CC52246DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-71-4
Molecular Formula C13H19NO2
Molecular Weight (g/mol) 221.3
MDL Number MFCD09064962
InChI Key ZTMRIJRRCHAUNA-UHFFFAOYSA-N
Synonym n-methyl-2-tetrahydropyran-4-yloxy benzylamine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl oxy phenyl methanamine, methyl 2-oxan-4-yloxy phenyl methyl amine, n-methyl-n-2-tetrahydro-2h-pyran-4-yloxy benzyl amine, n-methyl-1-2-oxan-4-yl oxy phenyl methanamine, methyl 2-2h-3,4,5,6-tetrahydropyran-4-yloxy phenyl methyl amine, n-methyl-1-2-tetrahydro-2h-pyran-4-yl ;oxy ;phenyl ;methanamine
PubChem CID 24229518
IUPAC Name N-methyl-1-[2-(oxan-4-yloxy)phenyl]methanamine
SMILES CNCC1=CC=CC=C1OC2CCOCC2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.3
Percent Purity 95%
Chemical Name or Material N-Methyl-2-(tetrahydropyran-4-yloxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.