missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(phenoxymethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 10143724
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10143724 1 g 1g
10012324 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143724 Supplier Thermo Scientific Maybridge Supplier No. CC58546DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-17-9
Molecular Formula C15H17NO
Molecular Weight (g/mol) 227.307
MDL Number MFCD09025894
InChI Key WFUPXVCKIKJBIM-UHFFFAOYSA-N
Synonym n-methyl-2-phenoxymethyl benzylamine, methyl 2-phenoxymethyl phenyl methyl amine, n-methyl-1-2-phenoxymethyl phenyl methanamine, benzenemethanamine,n-methyl-2-phenoxymethyl
PubChem CID 18525895
IUPAC Name N-methyl-1-[2-(phenoxymethyl)phenyl]methanamine
SMILES CNCC1=CC=CC=C1COC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 227.31
Percent Purity 97%
Chemical Name or Material N-methyl-2-(phenoxymethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.