missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-2-nitroaniline, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Methyl-2-nitroaniline |
| CAS | 612-28-2 |
| Molecular Formula | C7 H8 N2 O2 |
| Quantity | 5 g |
| Synonym | Benzenamine','N-methyl-2-nitro-, Aniline','N-methyl-o-nitro- (6CI,7CI,8CI), N-Methyl-2-nitrobenzenamine, (Methyl)(2-nitrophenyl)amine, 2-(Methylamino)nitrobenzene, 2-Nitro-N-methylaniline, 4-(N-Methylamino)-3-nitrobenzene, N-Methyl-2-nitroaniline, N-Methyl-o-nitroaniline, NSC 86672, o-(Methylamino)nitrobenzene, o-Nitro-N-methylaniline |
| InChI Formula | InChI=1S/C7H8N2O2/c1-8-6-4-2-3-5-7(6)9(10)11/h2-5,8H,1H3 |
| SMILES | CNc1ccccc1[N+](=O)[O-] |
| IUPAC Name | N-methyl-2-nitroaniline |
| Molecular Weight (g/mol) | 152.15 |
| Formula Weight | 152.0586 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction