missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(2-morpholinopyrimidin-5-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10113224
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 937796-14-0
Molecular Formula C10H16N4O
Molecular Weight (g/mol) 208.265
MDL Number MFCD09966161
InChI Key PFYPMSRZQMFCJO-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinopyrimidin-5-yl methylamine, methyl 2-morpholin-4-yl pyrimidin-5-yl methyl amine, methyl 2-morpholin-4-ylpyrimidin-5-yl methyl amine, n-methyl-1-2-morpholin-4-yl pyrimidin-5-yl methanamine
PubChem CID 42556099
IUPAC Name N-methyl-1-(2-morpholin-4-ylpyrimidin-5-yl)methanamine
SMILES CNCC1=CN=C(N=C1)N2CCOCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 208.26
Percent Purity 97%
Chemical Name or Material N-methyl-(2-morpholinopyrimidin-5-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'