missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2,2,2-trifluoroacetamide, Thermo Scientific™

Product Code. 15530524 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15530524 5 g 5g
15540524 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15530524 Supplier Thermo Scientific Acros Supplier No. 221570050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 815-06-5
Molecular Formula C3H4F3NO
Molecular Weight (g/mol) 127.07
InChI Key IQNHBUQSOSYAJU-UHFFFAOYSA-N
Synonym n-methyltrifluoroacetamide, n-methyl-2,2,2-trifluoroacetamide, acetamide, 2,2,2-trifluoro-n-methyl, 2,2,2-trifluoro-n-methyl-acetamide, n-metnyl-2,2,2-trifluoroacetamide, methyltrifluoroacetamide, acmc-209plm, n-methyl-trifluoroacetamide, n-methyltrifluoro-acetamide
PubChem CID 69948
IUPAC Name 2,2,2-trifluoro-N-methylacetamide
SMILES CNC(=O)C(F)(F)F

Specifications

Melting Point 48.0°C to 52.0°C
Boiling Point 156.0°C to 157.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CF3CONHCH3
Quantity 5 g
Formula Weight 127.07
Percent Purity ≥97.5% (GC)
Chemical Name or Material N-Methyl-2,2,2-trifluoroacetamide
compliance-icons
Product Identifier
  • N-Methyl-2,2,2-trifluoroacetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.