missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(1H-1,2,4-triazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 11360657
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-64-5
Molecular Formula C11H14N4
Molecular Weight (g/mol) 202.261
MDL Number MFCD09025848
InChI Key VHDVEBIJXKJALO-UHFFFAOYSA-N
Synonym n-methyl-2-1h-1,2,4-triazol-1-ylmethyl benzylamine, methyl 2-1,2,4-triazol-1-ylmethyl phenyl methyl amine, methyl 2-1,2,4-triazolylmethyl phenyl methyl amine, n-methyl-1-2-1h-1,2,4-triazol-1-yl methyl phenyl methanamine
PubChem CID 18525820
IUPAC Name N-methyl-1-[2-(1,2,4-triazol-1-ylmethyl)phenyl]methanamine
SMILES CNCC1=CC=CC=C1CN2C=NC=N2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 202.26
Percent Purity 97%
Chemical Name or Material N-methyl-2-(1H-1, 2, 4-triazol-1-ylmethyl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'