missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1-propylamine, 97%

Product Code. 11488726
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488726 25 g 25g
11468726 1 g 1g
11478726 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488726 Supplier Thermo Scientific Alfa Aesar Supplier No. A16903.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-35-0 | C4H11N | 73.139 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009361
InChI Key GVWISOJSERXQBM-UHFFFAOYSA-N
Synonym n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
PubChem CID 12315
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC

Specifications

Density 0.713
Boiling Point 62°C to 64°C
Flash Point −1.1°C (30°F)
Odor Amine-like
Quantity 25 g
UN Number UN2733
Formula Weight 73.14
Percent Purity 97%
Chemical Name or Material N-Methyl-1-propylamine
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Methyl-1-propylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: N-Methylpropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.