missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine, Thermo Scientific™

Catalog Number 10716925
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
1034566-07-8
C11H13N3
187.25
IZYNHWOVGJNYQF-UHFFFAOYSA-N
methyl 1-phenylimidazol-2-yl methyl amine, n-methyl-1-phenyl-1h-imidazol-2-yl methyl amine, n-methyl-1-1-phenyl-1h-imidazol-2-yl methanamine
45594302
This item is not returnable. View return policy
1034566-07-8
C11H13N3
187.25
IZYNHWOVGJNYQF-UHFFFAOYSA-N
methyl 1-phenylimidazol-2-yl methyl amine, n-methyl-1-phenyl-1h-imidazol-2-yl methyl amine, n-methyl-1-1-phenyl-1h-imidazol-2-yl methanamine
45594302
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 97%
Chemical Name or Material N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'