missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 187.24 |
Percent Purity | 97% |
Chemical Name or Material | N-methyl-[(1-phenyl-1H-imidazol-2-yl)methyl]amine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'