missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-(1-methylindolin-5-yl)methylamine, 90%, Thermo Scientific™

Product Code. 10692665
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10692665 1 g Amber Glass Bottle 1g
10376664 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10692665 Supplier Thermo Scientific Maybridge Supplier No. CC58746DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 906352-81-6
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD09817499
InChI Key QPUPWFNEDMKWGL-UHFFFAOYSA-N
Synonym 1-methyl-5-methylamino methyl indoline, 2,3-dihydro-1-methyl-5-methylamino methyl-1h-indole, methyl 1-methyl-2,3-dihydroindol-5-yl methyl amine, methyl 1-methyl-2,3-dihydro-1h-indol-5-yl methyl amine, methyl 1-methylindolin-5-yl methyl amine, n-methyl-1-1-methylindolin-5-yl methanamine, 1-2,3-dihydro-1-methyl-1h-indol-5-yl-n-methylmethylamine, n-methyl-1-1-methyl-2,3-dihydro-1h-indol-5-yl methanamine
PubChem CID 24229592
IUPAC Name N-methyl-1-(1-methyl-2,3-dihydroindol-5-yl)methanamine
SMILES CNCC1=CC2=C(C=C1)N(C)CC2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 176.26
Percent Purity 90%
Chemical Name or Material N-methyl-(1-methylindolin-5-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.