Learn More
n-methyl-(1-methyl-1h-indol-7-yl)methylamine, 90%, Thermo Scientific™
Brand: Thermo Scientific Maybridge CC66946DA
Code : 31
Additional Details : CAS Number : 709649-75-2 Weight : 0.05100kg Transport : UN number : 3259 Chem class : 8 Pack group : III
Packaging | Amber Glass Bottle |
---|---|
Quantity | 1g |
Chemical Identifiers
709649-75-2 | |
174.247 | |
JERMRTDADLXHSD-UHFFFAOYSA-N | |
33589520 | |
CNCC1=CC=CC2=C1N(C=C2)C |
C11H14N2 | |
MFCD11506353 | |
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine | |
N-methyl-1-(1-methylindol-7-yl)methanamine |
Specifications
N-methyl-(1-methyl-1H-indol-7-yl)methylamine | |
89.5 | |
90% | |
MFCD11506353 | |
methyl 1-methylindol-7-yl methyl amine, n-methyl-1-1-methyl-1h-indol-7-yl methylamine, n-methyl-1-methyl-1h-indol-7-yl methylamine, 1h-indole-7-methanamine, n,1-dimethyl, n-methyl-1-1-methyl-1h-indol-7-yl methanamine, 1h-indole-7-methanamine,n,1-dimethyl, methyl-1-methyl-1h-indol-7-ylmethyl amine, 7-methylamino methyl-1-methyl-1h-indole, methyl 1-methyl-1h-indol-7-yl methyl amine | |
JERMRTDADLXHSD-UHFFFAOYSA-N | |
N-methyl-1-(1-methylindol-7-yl)methanamine | |
33589520 | |
90% |
709649-75-2 | |
100.0 | |
C11H14N2 | |
Amber Glass Bottle | |
1g | |
CNCC1=CC=CC2=C1N(C=C2)C | |
174.247 | |
174.25 |