missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,3-propanediamine, 99%

Product Code. 11400937
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400937 500 g 500g
11490927 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11400937 Supplier Thermo Scientific Alfa Aesar Supplier No. B23319.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6291-84-5 | C4H12N2 | 88.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6291-84-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008209
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
PubChem CID 80511
IUPAC Name N'-methylpropane-1,3-diamine
SMILES CNCCCN

Specifications

Melting Point -72°C
Density 0.844
Boiling Point 139°C to 141°C
Flash Point 35°C (96°F)
Refractive Index 1.4475
Quantity 500 g
UN Number UN2734
Beilstein 878143
Sensitivity Air Sensitive
Formula Weight 88.15
Percent Purity 99%
Chemical Name or Material N-Methyl-1,3-propanediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.