missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,2-phenylenediamine, 97%

Product Code. 10618360 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10618360 5 mL Glass bottle 5mL
10311670 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10618360 Supplier Thermo Scientific Acros Supplier No. 434230050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4760-34-3 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4760-34-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00192314
InChI Key RPKCLSMBVQLWIN-UHFFFAOYSA-N
Synonym n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
PubChem CID 78498
IUPAC Name 2-N-methylbenzene-1,2-diamine
SMILES CNC1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 22°C
Color Brown to Yellow
Density 1.0750g/mL
Boiling Point 123°C to 124°C (10.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 1.075
Formula Weight 122.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Methyl-1, 2-phenylenediamine
Show More Show Less
compliance-icons
Product Identifier
  • N-Methyl-1,2-phenylenediamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.