missing translation for 'onlineSavingsMsg'
Learn More

N-Methoxy-N-methylacetamide, 98%

Product Code. 10581984 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10581984 25 g Glass bottle 25g
10191092 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10581984 Supplier Thermo Scientific Acros Supplier No. 369310250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78191-00-1 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78191-00-1
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.12
MDL Number MFCD00060098
InChI Key OYVXVLSZQHSNDK-UHFFFAOYSA-N
Synonym n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
PubChem CID 537505
IUPAC Name N-methoxy-N-methylacetamide
SMILES CON(C)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.9700g/mL
Boiling Point 152°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CON(OCH3)CH3
Refractive Index 1.4240 to 1.426
Quantity 25 g
Specific Gravity 0.97
Solubility Information Solubility in water: soluble
Formula Weight 103.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N-Methoxy-N-methylacetamide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • N-Methoxy-N-methylacetamide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.