missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine, 98%, MP Biomedicals™

Product Code. 11451821 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11451821 100 g 100g
11461821 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11451821 Supplier MP Biomedicals Supplier No. 0219028980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 137-16-6
Molecular Formula C15H28NNaO3
Molecular Weight (g/mol) 293.38
MDL Number MFCD00042728
InChI Key KSAVQLQVUXSOCR-UHFFFAOYSA-M
Synonym sarkosyl nl, sodium lauroyl sarcosinate, n-lauroylsarcosine sodium salt, sodium n-lauroylsarcosinate, sodium lauroylsarcosinate, sarcosyl nl, maprosyl 30, compound 105, gardol, hamposyl l-30
PubChem CID 23668817
IUPAC Name sodium;2-[dodecanoyl(methyl)amino]acetate
SMILES [Na+].CCCCCCCCCCCC(=O)N(C)CC([O-])=O

Specifications

Color White
Assay Percent Range 0.98
Quantity 100 g
Formula Weight 293.4
Physical Form Powder
Chemical Name or Material N-Lauroylsarcosine
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • N-Lauroylsarcosine Sodium Salt
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Mention de danger
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Conseils de prudence
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P320-Specific treatment is urgent (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Informations supplémentaires
  • MIXTURE LIST-Contain: N-Lauroylsarcosine Sodium Salt
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.