missing translation for 'onlineSavingsMsg'
Learn More

N-Lauroylsarcosine, 98.7%, MP Biomedicals™

Product Code. 11438232 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 g
100 g
Unit Size:
100g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438232 100 g 100g
17871381 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438232 Supplier MP Biomedicals Supplier No. 0219011080
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 97-78-9
Molecular Formula C15H29NO3
Molecular Weight (g/mol) 271.40
MDL Number MFCD00021749
InChI Key BACYUWVYYTXETD-UHFFFAOYSA-N
Synonym Hamposyl L
PubChem CID 7348
IUPAC Name 2-(N-methyldodecanamido)acetic acid
SMILES CCCCCCCCCCCC(=O)N(C)CC(O)=O

Specifications

Color White
Physical Form White, waxy solid
Quantity 100 g
Assay Percent Range 98.7%
Formula Weight 271.4
Chemical Name or Material N-Lauroylsarcosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.