missing translation for 'onlineSavingsMsg'
Learn More

N-L-Gamma-GLUTAMYL-L-LEUCINE, TRC

Product Code. 30240927
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240927 10 mg 10mg
30240506 50 mg 50mg
30240735 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240927 Supplier Toronto Research Chemicals Supplier No. TRCG24514010MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2566-39-4
Molecular Formula C11H20N2O5
Molecular Weight (g/mol) 260.29
Synonym (S)-2-Amino-5-((S)-1-carboxy-3-methylbutylamino)-5- oxopentanoic acid
IUPAC Name (2S)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)CC[C@H](N)C(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C11H20N2O5/c1-6(2)5-8(11(17)18)13-9(14)4-3-7(12)10(15)16/h6-8H,3-5,12H2,1-2H3,(H,13,14)(H,15,16)(H,17,18)/t7-,8-/m0/s1
Quantity 10 mg
Formula Weight 260.1372
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material N-L-gamma-GLUTAMYL-L-LEUCINE
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.