Learn More
N-Isopropylmethylamine, 98%, Thermo Scientific Chemicals
Description
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N-Isopropylmethylamine is used in the generation of in situ micelles having distinct sizes, by sequential self-assembly of a series of stimuli-responsive amphiphilic block copolymers, prepared by reversible addition-fragmentation chain transfer (RAFT) polymerization. It is suitable for use in the synthesis of novel 5-ethoxy-N,N-dialkyl-[α,α,β,β-H4]-tryptamines and their [α,α,β,β,β-D4]-counterparts following the Speeter and Anthony procedure.
Solubility
Soluble in chloroform, DMSO, methanol.
Notes
Air Sensitive. Store away from air. Store under dry inert gas. Do not store together with acids. Incompatible with acids , air and oxidizing agents.
Specifications
Color | Colorless |
Density | 0.702 g/mL |
Boiling Point | 50°C to 53°C |
Flash Point | -32°C (-26°F) |
Linear Formula | (CH3)2CHNHCH3 |
Refractive Index | 1.348 |
Quantity | 25 g |
UN Number | UN2734 |
Beilstein | 1730877 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only