missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropylbenzylamine, TRC

Product Code. 30401221
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401221 1 g -
30401916 25 g 25g
30399916 100 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401221 Supplier Toronto Research Chemicals Supplier No. TRCI8243551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-97-6
Molecular Formula C10 H15 N
Molecular Weight (g/mol) 149.23
Synonym N-(1-Methylethyl)benzenemethanamine, (1-Methylethyl)(phenylmethyl)amine, Benzylisopropylamine, N-Benzyl-N-isopropylamine, N-Isopropyl-N-benzylamine, NSC 60295
IUPAC Name N-benzylpropan-2-amine
SMILES CC(C)NCc1ccccc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H15N/c1-9(2)11-8-10-6-4-3-5-7-10/h3-7,9,11H,8H2,1-2H3
Formula Weight 149.1204
Chemical Name or Material N-Isopropylbenzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.