missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl Noratropine, TRC

Product Code. 30399873
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399873 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30399873 Supplier Toronto Research Chemicals Supplier No. TRCI87268010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Isopropyl Noratropine
CAS 22235-81-0
Purity Grade Notes HPLC
Molecular Formula C19 H27 N O3
Quantity 10 mg
Synonym (1R,3r,5S)-8-(1-Methylethyl)-8-aza-bicyclo[3.2.1]oct-3-yl (2RS)-3-hydroxy-2-phenyl-propanoate, 1αH,5αH-Nortropan-3α-ol, 8-isopropyl-, (±)-tropate (ester) (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, 8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, endo-(±)-, Benzeneacetic acid, α-(hydroxymethyl)-, 8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, endo-, N-Isopropylnoratropine, Tropic acid, 8-isopropyl-3α-nortropanyl ester, Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-(1-methylethyl)-8-azabicyclo[3.2.1]oct-3-yl ester, Ipratropium Bromide Imp E (EP)
InChI Formula InChI=1S/C19H27NO3/c1-13(2)20-15-8-9-16(20)11-17(10-15)23-19(22)18(12-21)14-6-4-3-5-7-14/h3-7,13,15-18,21H,8-12H2,1-2H3/t15-,16+,17+,18-/m0/s1
SMILES CC(C)N1[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3
IUPAC Name [(1S,5R)-8-propan-2-yl-8-azabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate
Molecular Weight (g/mol) 317.42
Formula Weight 317.1991
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.