missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-N'-phenylbenzene-1,4-diamine, TRC

Product Code. 30138700
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138700 10 g 10g
30010704 250 mg 250mg
30011635 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138700 Supplier Toronto Research Chemicals Supplier No. TRCN24774510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-72-4
Molecular Formula C15 H18 N2
Molecular Weight (g/mol) 226.32
Synonym 1,4-Benzenediamine, N-(1-methylethyl)-N'-phenyl- (9CI), p-Phenylenediamine, N-isopropyl-N'-phenyl- (6CI,7CI,8CI), N1-(1-Methylethyl)-N4-phenyl-1,4-benzenediamine, 1-N-Phenyl-4-N-(propan-2-yl)benzene-1,4-diamine, 1-N-Phenyl-4-N-propan-2-ylbenzene-1,4-diamine, 4-(Isopropylamino)diphenylamine, 4-Anilino-N-isopropylaniline, 4-Isopropylamino-1-phenylaminobenzene, 4010A, 4010NA, 810NA, AO-CD, ASM 4010MA, Accinox ZA, Antage 3C, Antigen 3C, Antigene 3C, Antioxidant 3C, Antioxidant 4010A, Antioxidant 4010NA, Antioxidant 40NA, Antioxidant CD, Antioxidant IP, Antioxidant IPPD, Antiozonant PD 1, Cyzone IP, Diafen FP, Diafen PP, Diaphen FP, Diaphene FP, Dusantox IPPD, Flexzone 3C, Flexzone 3P, IPPD, Ipognox 44, N 1040, N-(1-Methylethyl)-N'-phenyl-1,4-benzenediamine, N-2-Propyl-N'-phenyl-p-phenylenediamine, N-Isopropyl-N'-phenyl-1,4-benzenediamine, N-Isopropyl-N'-phenyl-1,4-phenylenediamine, N-Isopropyl-N'-phenyl-4-phenylenediamine, N-Isopropyl-N'-phenyl-p-phenyldiamine, N-Isopropyl-N'-phenyl-p-phenylendiamine, N-Isopropyl-N'-phenyl-p-phenylenediamine, N-Phenyl-N1-isopropyl-p-phenylenediamine, N-Phenyl-N'-isopropyl-1,4-phenylenediamine, N-Phenyl-N'-isopropyl-p-phenylanediamine, N-Phenyl-N'-isopropyl-p-phenylenediamine, NA 4010, NSC 41029, Naugard I 4, Nocrac 3C, Nocrac 810MA, Nocrac 810NA, Nonox ZA, Orflex PP, Ozonone 3C, PD 1, Permanax 115, Permanax IPPD, Santoflex 36, Santoflex IP, Santoflex IPPD, Vulkanox 4010, Vulkanox 4010A, Vulkanox 4010NA, Vulkanox 4010NA-LG, Vulkanox 4020NA, p-(Isopropylamino)diphenylamine, N-Isopropyl-N'-phenyl-p-phenylenediamine (IPPD)
IUPAC Name 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
SMILES CC(C)Nc1ccc(Nc2ccccc2)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H18N2/c1-12(2)16-14-8-10-15(11-9-14)17-13-6-4-3-5-7-13/h3-12,16-17H,1-2H3
Formula Weight 226.147
Percent Purity >95%
Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine (IPPD)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.