missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-N'-phenyl-p-phenylenediamine, 95%

Product Code. 11477056
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477056 250 g 250g
11467056 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477056 Supplier Thermo Scientific Alfa Aesar Supplier No. B25378.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 101-72-4 | C15H18N2 | 226.323 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-72-4
Molecular Formula C15H18N2
Molecular Weight (g/mol) 226.323
MDL Number MFCD00026348
InChI Key OUBMGJOQLXMSNT-UHFFFAOYSA-N
Synonym ippd, n-isopropyl-n'-phenyl-p-phenylenediamine, n-isopropyl-n'-phenyl-1,4-phenylenediamine, cyzone, 4-anilino-n-isopropylaniline, antigen 3c, antioxidant ip, santoflex ip, diaphen fp, cyzone ip
PubChem CID 7573
ChEBI CHEBI:63569
IUPAC Name 1-N-phenyl-4-N-propan-2-ylbenzene-1,4-diamine
SMILES CC(C)NC1=CC=C(C=C1)NC2=CC=CC=C2

Specifications

Melting Point 74°C to 80°C
Density 1.1
Boiling Point 161°C (1 mmHg)
Flash Point 172°C (342°F)
Quantity 250 g
UN Number UN3077
Sensitivity Air Sensitive
Formula Weight 226.32
Percent Purity 95%
Chemical Name or Material N-Isopropyl-N'-phenyl-p-phenylenediamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.