missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-N-methylamine, TRC

Product Code. 30136157
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136157 10 g 10g
30006494 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136157 Supplier Toronto Research Chemicals Supplier No. TRCI87225010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4747-21-1
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.14
Synonym 2-(Methylamino)propane, Isopropylmethylamine, N,1-dimethyl-Ethylamine, N-methyl-Isopropylamine, Methylisopropylamine, N-Methyl-N-(1-methylethyl)amine, N-Methylisopropylamine, NSC 1065;
IUPAC Name N-methylpropan-2-amine
SMILES CNC(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H11N/c1-4(2)5-3/h4-5H,1-3H3
Formula Weight 73.0891
Chemical Name or Material N-Isopropyl-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.