missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl Carvedilol, TRC

Product Code. 30390017
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390017 10 mg 10mg
30393846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390017 Supplier Toronto Research Chemicals Supplier No. TRCI82439010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246819-01-1
Molecular Formula C27 H32 N2 O4
Molecular Weight (g/mol) 448.55
Synonym 2-Propanol, 1-(9H-carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl](1-methylethyl)amino]-, 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl](1-methylethyl)amino]-2-propanol, 1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl](1-methylethyl)amino]propan-2-ol, 1-(9H-Carbazol-4-yloxy)-3-[isopropyl[2-(2-methoxyphenoxy)ethyl]amino]propan-2-ol, N-Isopropylcarvedilol, Carvedilol RC F (USP)
IUPAC Name 1-(9H-carbazol-4-yloxy)-3-[2-(2-methoxyphenoxy)ethyl-propan-2-ylamino]propan-2-ol
SMILES COc1ccccc1OCCN(CC(O)COc2cccc3[nH]c4ccccc4c23)C(C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C27H32N2O4/c1-19(2)29(15-16-32-25-13-7-6-12-24(25)31-3)17-20(30)18-33-26-14-8-11-23-27(26)21-9-4-5-10-22(21)28-23/h4-14,19-20,28,30H,15-18H2,1-3H3
Formula Weight 448.2362
Chemical Name or Material N-Isopropyl Carvedilol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.