missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-4-nitroaniline, 97+%, Thermo Scientific™

Product Code. 15499067
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15499067 250 mg 250mg
15479067 1 g 1g
15489067 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15499067 Supplier Thermo Scientific Alfa Aesar Supplier No. H52375.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25186-43-0
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00128997
InChI Key VTSUWHFLMJLYKN-UHFFFAOYSA-N
Synonym n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro
PubChem CID 32833
IUPAC Name 4-nitro-N-propan-2-ylaniline
SMILES CC(C)NC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Quantity 250 mg
UN Number UN2811
Formula Weight 180.2
Percent Purity ≥97%
Chemical Name or Material N-Isopropyl-4-nitroaniline
compliance-icons
Product Identifier
  • N-Isopropyl-4-nitroaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: N-(1-methylethyl)-4-nitrobenzenamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.