missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-2-methyl-2-propyl-3-hydroxypropyl Carbamate, TRC

Product Code. 30137316
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137316 500 mg 500mg
30006709 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137316 Supplier Toronto Research Chemicals Supplier No. TRCI872625500MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25462-17-3
Molecular Formula C11 H23 N O3
Molecular Weight (g/mol) 217.31
Synonym (2RS)-2-(Hydroxymethyl)-2-methylpentyl (1-Methylethyl)carbamate, Carisoprodol Imp. A (EP)
IUPAC Name [2-(hydroxymethyl)-2-methylpentyl] N-propan-2-ylcarbamate
SMILES CCCC(C)(CO)COC(=O)NC(C)C

Specifications

InChI Formula InChI=1S/C11H23NO3/c1-5-6-11(4,7-13)8-15-10(14)12-9(2)3/h9,13H,5-8H2,1-4H3,(H,12,14)
Quantity 500 mg
Formula Weight 217.1678
Shipping Condition Room Temperature
Chemical Name or Material (2RS)-2-(Hydroxymethyl)-2-methylpentyl (1-Methylethyl)carbamate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.