missing translation for 'onlineSavingsMsg'
Learn More

N-Iodosuccinimide, 97%

Product Code. 11476867
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476867 25 g 25g
11466867 5 g 5g
11486867 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476867 Supplier Thermo Scientific Alfa Aesar Supplier No. A14320.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 516-12-1 | C4H4INO2 | 224.99 g/mol

N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Iodosuccinimide is used in the preparation of vinyl sulfones from olefins and benzenesulfinic acid. It acts as a source for I+ and involved in Hunsdiecker reactions for the conversion of cinnamic acids, and propiolic acids to the corresponding alfa-halostyrenes and 1-halo-1-alkynes respectively. It is also used to hydrolyze thioglycosides to 1-hydroxyglycosides with triluoroacetic acid. It is involved in the preparation of iodobenzene from 1,6-diynes. Further, it acts as an iodinating agent in chemical synthesis.

Solubility
Soluble in dioxane, tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-12-1
Molecular Formula C4H4INO2
Molecular Weight (g/mol) 224.99
MDL Number MFCD00005512
InChI Key LQZMLBORDGWNPD-UHFFFAOYSA-N
Synonym n-iodosuccinimide, succiniodimide, n-iodo succinimide, 2,5-pyrrolidinedione, 1-iodo, unii-3cos3x3n4p, nis, succinimide, n-iodo, 3cos3x3n4p, 1-iodo-pyrrolidine-2,5-dione, 1-iodoazolidine-2,5-dione
PubChem CID 120273
ChEBI CHEBI:53204
IUPAC Name 1-iodopyrrolidine-2,5-dione
SMILES IN1C(=O)CCC1=O

Specifications

Melting Point ∽196°C (decomposition)
Density 2.245
Assay Percent Range 97%
Quantity 25 g
Beilstein 113917
Sensitivity Light and moisture sensitive
Solubility Information Soluble in dioxane,tetrahydrfuran and acetonitrile. Insoluble in ether and carbon tetrachloride.
Formula Weight 224.99
Chemical Name or Material N-Iodosuccinimide
compliance-icons
Product Identifier
  • N-Iodosuccinimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-iodo-2,5-Pyrrolidinedione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.