missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysulfosuccinimide sodium salt, 95%

Product Code. p-4228730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10616260 1 g Glass bottle 1g
10666450 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10616260 Supplier Thermo Scientific Acros Supplier No. 438650010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Coupling reagent | CAS: 106627-54-7 | C4H4NNaO6S | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106627-54-7
Molecular Formula C4H4NNaO6S
Molecular Weight (g/mol) 217.13
InChI Key RPENMORRBUTCPR-UHFFFAOYSA-M
Synonym n-hydroxysulfosuccinimide sodium salt, sulfo-nhs, sodium 1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate, n-hydroxysulfosuccinimidesodiumsalt, 3-pyrrolidinesulfonic acid, 1-hydroxy-2,5-dioxo-, monosodium salt, sodium 1-hydroxy-2,5-dioxo-pyrrolidine-3-sulfonate, n-hydroxusulfosuccinimide sodium, n-hydroxy-2-sodiosulfo succinimide, n-hydroxysulfosuccinimide,sodium salt, n-hydroxusulfosuccinimide sodium salt
PubChem CID 3520574
IUPAC Name sodium;1-hydroxy-2,5-dioxopyrrolidine-3-sulfonate
SMILES C1C(C(=O)N(C1=O)O)S(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 250.0°C
Color White
Elemental Analysis 21.46 to 23.02% C, 6.19 to 6.71% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Formula Weight 217.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N-Hydroxysulfosuccinimide sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • N-Hydroxysulfosuccinimide Sodium Salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.