missing translation for 'onlineSavingsMsg'
Learn More

N-Hydroxysuccinimide, TRC

Product Code. 30008229
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008229 25 g 25g
30007360 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008229 Supplier Toronto Research Chemicals Supplier No. TRCH95358525G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6066-82-6
Molecular Formula C4 H5 N O3
Molecular Weight (g/mol) 115.09
IUPAC Name 1-hydroxypyrrolidine-2,5-dione
SMILES ON1C(=O)CCC1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H5NO3/c6-3-1-2-4(7)5(3)8/h8H,1-2H2
Formula Weight 115.0269
Percent Purity >95%
Chemical Name or Material N-Hydroxysuccinimide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.